Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPTG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032520.4 | NP_115909.1 | 10 | 822 | ||
XM_017023782.1 | XP_016879271.1 | 10 | 838 | |||
XM_017023783.1 | XP_016879272.1 | 8 | 710 | |||
GenBank mRNA | AF302786.1 | - | 10 | 755 | ||
AK126110.1 | - | 4 | 1978 | |||
AK312067.1 | - | 10 | 790 | |||
AY203933.1 | - | 10 | 753 | |||
BC014592.1 | - | 10 | 782 | 4824054 | ||
BC131525.1 | - | 9 | 717 |