Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318970.1 | NP_001305899.1 | 5 | 367 | ||
NM_001623.4 | NP_001614.3 | 5 | 428 | |||
NM_032955.2 | NP_116573.1 | 2 | 257 | |||
XM_005248870.4 | XP_005248927.1 | 4 | 618 | |||
XM_017010332.1 | XP_016865821.1 | 4 | 562 | |||
GenBank mRNA | AF088038.1 | - | 5 | 374 | ||
AF299327.1 | - | 2 | 145 | |||
AK290771.1 | - | 5 | 363 | |||
AV716806.1 | - | 5 | 348 | |||
AV735626.1 | - | 5 | 473 | |||
BC009474.1 | - | 5 | 407 | 4249306 | ||
CD249922.1 | - | 2 | 293 | |||
CD521003.1 | - | 5 | 335 | |||
CR542153.1 | - | 2 | 145 | |||
D86438.1 | - | 5 | 316 | |||
EF070981.1 | - | 2 | 150 | |||
EF070982.1 | - | 1 | 365 | |||
EF070983.1 | - | 5 | 307 | |||
JQ782414.1 | - | 5 | 402 | |||
JQ782415.1 | - | 5 | 341 | |||
U19713.1 | - | 5 | 331 | |||
U49392.1 | - | 5 | 392 | |||
U95213.1 | - | 3 | 796 |