Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282129.1 | NP_001269058.1 | 16 | 3280 | ||
NM_033389.3 | NP_203747.2 | 15 | 3011 | |||
XM_005258058.3 | XP_005258115.1 | 17 | 3173 | |||
XM_005258059.2 | XP_005258116.1 | 14 | 3147 | |||
XM_005258060.3 | XP_005258117.1 | 3 | 1740 | |||
XM_006722149.3 | XP_006722212.1 | 14 | 6778 | |||
XM_011525402.2 | XP_011523704.1 | 16 | 3008 | |||
XM_011525404.2 | XP_011523706.1 | 14 | 3771 | |||
XM_011525405.2 | XP_011523707.1 | 14 | 3819 | |||
XM_011525406.2 | XP_011523708.1 | 13 | 3098 | |||
XM_011525407.1 | XP_011523709.1 | 13 | 2843 | |||
XM_017025251.1 | XP_016880740.1 | 16 | 3216 | |||
XM_017025252.1 | XP_016880741.1 | 14 | 3077 | |||
GenBank mRNA | AB051512.1 | - | 3 | 1829 | ||
AB099290.1 | - | 15 | 2971 | |||
AK091432.1 | - | 2 | 1400 | |||
AK296413.1 | - | 16 | 3145 |