Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052838.4 | NP_443070.5 | 3 | 204 | ||
XM_017022991.1 | XP_016878480.1 | 3 | 184 | |||
GenBank mRNA | AF085235.1 | - | 2 | 38 | ||
AK301045.1 | - | 2 | 36 | |||
AK303390.1 | - | 3 | 167 | |||
AY034176.1 | - | 2 | 38 | |||
BC012161.1 | - | 1 | 26 | 4576220 | ||
BQ072902.1 | - | 3 | 217 |