Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRPM3 SIRNA Assays ›
Gene Symbol:
TRPM3
Gene Name:
transient receptor potential cation channel subfamily M member 3
Gene Aliases:
GON-2, LTRPC3, MLSN2
Chromosome Location:
Chr.9: 70529063 - 71446989 on Build GRCh38
UniGene ID:
Hs.47288
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
29003
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRPM3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007470.1
NP_001007471.1
5
612
NM_001007471.2
NP_001007472.2
5
960
NM_020952.4
NP_066003.3
5
612
NM_024971.5
NP_079247.5
5
612
NM_206944.3
NP_996827.3
5
612
NM_206945.3
NP_996828.3
5
612
NM_206946.3
NP_996829.3
5
612
NM_206947.3
NP_996830.3
5
612
NM_206948.2
NP_996831.1
5
612
XM_011519035.2
XP_011517337.1
5
837
XM_011519036.2
XP_011517338.1
5
3762
XM_011519037.2
XP_011517339.1
5
3748
XM_011519038.2
XP_011517340.1
5
3745
XM_011519039.2
XP_011517341.1
5
3746
XM_011519040.2
XP_011517342.1
5
843
XM_011519041.2
XP_011517343.1
5
3745
XM_011519042.2
XP_011517344.1
5
843
XM_011519043.2
XP_011517345.1
5
876
XM_011519044.2
XP_011517346.1
5
3746
XM_011519045.2
XP_011517347.1
5
837
XM_011519046.2
XP_011517348.1
5
2540
XM_011519047.2
XP_011517349.1
5
2510
XM_017015141.1
XP_016870630.1
5
837
XM_017015142.1
XP_016870631.1
5
837
XM_017015143.1
XP_016870632.1
5
837
XM_017015144.1
XP_016870633.1
5
603
XM_017015145.1
XP_016870634.1
5
1281
XM_017015146.1
XP_016870635.1
5
679
XM_017015147.1
XP_016870636.1
5
2714
XM_017015148.1
XP_016870637.1
5
837
XM_017015149.1
XP_016870638.1
5
837
XM_017015150.1
XP_016870639.1
5
837
XM_017015151.1
XP_016870640.1
5
855
XM_017015152.1
XP_016870641.1
5
2515
XM_017015153.1
XP_016870642.1
5
847
XM_017015154.1
XP_016870643.1
5
983
XM_017015155.1
XP_016870644.1
5
2512
XM_017015156.1
XP_016870645.1
1
35
XM_017015157.1
XP_016870646.1
5
2510
XM_017015158.1
XP_016870647.1
5
2508
XM_017015159.1
XP_016870648.1
5
603
XM_017015160.1
XP_016870649.1
5
693
XM_017015161.1
XP_016870650.1
5
837
GenBank mRNA
AB099661.1
-
2
257
AB099662.1
-
2
257
AB099663.1
-
2
257
AB099664.1
-
2
257
AB099665.1
-
2
257
AF325212.1
-
5
583
AF536748.1
-
2
257
AF536749.1
-
2
257
AF536750.1
-
2
257
AF536751.1
-
2
257
AF536752.1
-
2
257
AF536753.1
-
2
257
AJ505025.1
-
5
960
AJ505026.1
-
5
960
AK021788.1
-
5
603
AK225732.1
-
5
603
AK308682.1
-
5
788
AL136545.1
-
5
793
BC022454.2
-
4
542
BC067733.1
-
5
574
BC094699.1
-
5
606
BC121821.1
-
3
458
BC134414.1
-
3
461
BC142972.1
-
5
629
KF987075.1
-
5
668
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
transporter
Gene Ontology Categories:
Function(s)
Process(es)
cation transport
detection of temperature stimulus
sensory perception of temperature stimulus
protein tetramerization
calcium ion transmembrane transport
calcium activated cation channel activity
cation channel activity
calcium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0