Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243773.1 | NP_001230702.1 | 4 | 3256 | ||
NM_001243774.1 | NP_001230703.1 | 3 | 3219 | |||
NM_053064.4 | NP_444292.1 | 4 | 3260 | |||
XM_006720173.2 | XP_006720236.1 | 4 | 3021 | |||
XM_011536846.2 | XP_011535148.1 | 4 | 2798 | |||
XM_017021376.1 | XP_016876865.1 | 5 | 3042 | |||
XM_017021377.1 | XP_016876866.1 | 6 | 3499 | |||
GenBank mRNA | AK026424.1 | - | 1 | 1604 | ||
AK026425.1 | - | 1 | 1604 | |||
AK092371.1 | - | 3 | 3113 | |||
AL832878.1 | - | 4 | 2899 | |||
BC060856.1 | - | 3 | 2798 | 5276390 |