Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014203.2 | NP_055018.2 | 12 | 1698 | ||
NM_130787.2 | NP_570603.2 | 12 | 1698 | |||
XM_011526556.2 | XP_011524858.1 | 12 | 2308 | |||
XM_011526557.2 | XP_011524859.1 | 12 | 2303 | |||
GenBank mRNA | AK094317.1 | - | 2 | 134 | ||
AL136925.1 | - | 12 | 1564 | |||
BC014214.2 | - | 12 | 1596 | 4111165 |