Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138983.2 | NP_620450.2 | 1 | 1472 | ||
GenBank mRNA | AL355743.1 | - | 1 | 616 | ||
AL834450.1 | - | 1 | 1465 | |||
BC026989.1 | - | 1 | 1349 | 4944525 | ||
BC033290.1 | - | 1 | 1479 | 4940343 |