Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006610.3 | NP_006601.2 | 3 | 348 | ||
NM_139208.2 | NP_631947.1 | 3 | 348 | |||
XM_017000097.1 | XP_016855586.1 | 3 | 348 | |||
GenBank mRNA | AB008047.1 | - | 3 | 342 | ||
AK290823.1 | - | 3 | 348 | |||
BC080556.1 | - | 3 | 336 | |||
DA638135.1 | - | 3 | 348 | |||
X98400.1 | - | 3 | 352 | |||
Y09926.1 | - | 3 | 332 | |||
Y18281.1 | - | 3 | 337 | |||
Y18282.1 | - | 2 | 286 | |||
Y18283.1 | - | 3 | 331 | |||
Y18284.1 | - | 3 | 337 |