Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267551.1 | NP_001254480.1 | 7 | 735 | ||
NM_001267552.1 | NP_001254481.1 | 8 | 832 | |||
NM_007169.2 | NP_009100.2 | 7 | 793 | |||
NM_148172.2 | NP_680477.1 | 7 | 801 | |||
NM_148173.1 | NP_680478.1 | 7 | 680 | |||
XM_006721418.3 | XP_006721481.2 | 7 | 682 | |||
XM_017024016.1 | XP_016879505.1 | 5 | 588 | |||
GenBank mRNA | AB029821.1 | - | 7 | 793 | ||
AF044214.1 | - | 7 | 766 | |||
AF113126.1 | - | 7 | 770 | |||
AF176806.1 | - | 7 | 743 | |||
AF176807.1 | - | 7 | 688 | |||
AK302251.1 | - | 8 | 837 | |||
BC000557.2 | - | 7 | 777 | 3162576 | ||
BC050593.1 | - | 7 | 778 | 6148983 |