Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTF3L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136497.2 | NP_001129969.1 | 5 | 676 | ||
NM_001243767.1 | NP_001230696.1 | 5 | 541 | |||
NM_152265.4 | NP_689478.1 | 6 | 743 | |||
GenBank mRNA | AK027582.1 | - | 5 | 557 | ||
AK027750.1 | - | 6 | 625 | |||
BC021004.1 | - | 5 | 520 | |||
BC022371.1 | - | 6 | 605 | 4739327 | ||
BC070378.1 | - | 6 | 633 | 6495112 | ||
BU785869.1 | - | 5 | 385 | |||
DA369498.1 | - | 5 | 462 |