Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HIF3A SIRNA Assays ›
Gene Symbol:
HIF3A
Gene Name:
hypoxia inducible factor 3 alpha subunit
Gene Aliases:
HIF-3A, HIF3-alpha-1, IPAS, MOP7, PASD7, bHLHe17
Chromosome Location:
Chr.19: 46297046 - 46343433 on Build GRCh38
UniGene ID:
Hs.420830
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
290127
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HIF3A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022462.4
NP_071907.4
3
358
NM_152794.3
NP_690007.1
4
454
NM_152795.3
NP_690008.2
4
422
NM_152796.2
NP_690009.1
3
358
XM_005259152.4
XP_005259209.1
5
550
XM_005259153.3
XP_005259210.2
2
221
XM_005259155.4
XP_005259212.1
5
550
XM_005259156.4
XP_005259213.1
5
550
XM_017027132.1
XP_016882621.1
5
550
XM_017027133.1
XP_016882622.1
5
550
XM_017027134.1
XP_016882623.1
4
420
XM_017027135.1
XP_016882624.1
4
454
XM_017027136.1
XP_016882625.1
5
550
XM_017027137.1
XP_016882626.1
4
454
XM_017027138.1
XP_016882627.1
4
420
XM_017027139.1
XP_016882628.1
3
344
XM_017027140.1
XP_016882629.1
3
274
XM_017027141.1
XP_016882630.1
3
316
XM_017027142.1
XP_016882631.1
3
344
GenBank mRNA
AB054067.1
-
4
385
AB118749.1
-
4
403
AB295039.1
-
4
403
AK021421.1
-
3
308
AK021653.1
-
4
448
AK024095.1
-
668
AK027725.1
-
4
403
AK297828.1
-
3
344
AK299699.1
-
2
268
AK304747.1
-
4
486
AK314680.1
-
4
454
AL541807.3
-
4
425
BC026308.1
-
4
413
4811824
BC080551.1
-
4
393
6250259
BI772109.1
-
4
479
CF593418.1
-
3
294
DA559979.1
-
3
344
DA562618.1
-
4
434
DB176660.1
-
4
422
DC423791.1
-
4
504
DR001068.1
-
4
430
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
transcription from RNA polymerase II promoter
apoptotic process
positive regulation of transcription from RNA polymerase II promoter
regulation of transcription from RNA polymerase II promoter in response to hypoxia
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-cp9cp:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline