Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146261.2 | NP_001139733.1 | 7 | 1123 | ||
NM_001146262.2 | NP_001139734.1 | 6 | 1002 | |||
NM_001146264.2 | NP_001139736.1 | 7 | 1123 | |||
NM_001256006.1 | NP_001242935.1 | 7 | 1207 | |||
NM_153262.3 | NP_694994.2 | 6 | 1002 | |||
XM_006711262.3 | XP_006711325.1 | 5 | 2181 | |||
XM_017000931.1 | XP_016856420.1 | 5 | 2181 | |||
XM_017000932.1 | XP_016856421.1 | 7 | 1197 | |||
XM_017000933.1 | XP_016856422.1 | 7 | 1060 | |||
XM_017000934.1 | XP_016856423.1 | 4 | 856 | |||
XM_017000935.1 | XP_016856424.1 | 5 | 739 | |||
GenBank mRNA | AB102948.1 | - | 5 | 911 | ||
AJ617623.1 | - | 6 | 945 | |||
AJ617624.1 | - | 6 | 1088 | |||
AJ617626.1 | - | 3 | 678 | |||
AJ617627.1 | - | 2 | 437 | |||
AK091517.1 | - | 6 | 925 | |||
BC130323.1 | - | 7 | 1056 | 40146354 | ||
BC144154.1 | - | 7 | 1056 | |||
BC144155.1 | - | 6 | 935 | |||
BC144157.1 | - | 7 | 1056 |