Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC2A14 SIRNA Assays ›
Gene Symbol:
SLC2A14
Gene Name:
solute carrier family 2 member 14
Gene Aliases:
GLUT14, SLC2A3P3
Chromosome Location:
Chr.12: 7812512 - 7891196 on Build GRCh38
UniGene ID:
Hs.655169
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
290136
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC2A14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286233.1
NP_001273162.1
16
2652
NM_001286234.1
NP_001273163.1
11
1898
NM_001286235.1
NP_001273164.1
13
2196
NM_001286236.1
NP_001273165.1
10
1737
NM_001286237.1
NP_001273166.1
10
1791
NM_153449.3
NP_703150.1
12
1934
XM_005253315.3
XP_005253372.1
11
1732
XM_005253317.4
XP_005253374.1
11
2251
XM_011520562.1
XP_011518864.1
14
2086
XM_011520563.2
XP_011518865.1
11
1746
XM_011520564.2
XP_011518866.1
10
1555
XM_011520565.2
XP_011518867.1
9
1584
XM_017018841.1
XP_016874330.1
12
1831
XM_017018842.1
XP_016874331.1
15
2402
XM_017018843.1
XP_016874332.1
15
2726
XM_017018844.1
XP_016874333.1
12
1781
XM_017018845.1
XP_016874334.1
12
2323
XM_017018846.1
XP_016874335.1
12
1907
XM_017018847.1
XP_016874336.1
11
1864
GenBank mRNA
AF481878.1
-
11
1717
AF481879.1
-
12
1794
AK122783.1
-
13
1983
AK126026.1
-
16
2433
AK299037.1
-
10
1742
AK302881.1
-
10
1549
AK316238.1
-
10
1742
AL110298.1
-
5
977
AL832448.1
-
9
1557
AY357941.1
-
11
1882
BC060766.1
-
11
1745
5297510
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
spermatogenesis
cell differentiation
glucose transmembrane transport
glucose transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0