Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002274.3 | NP_002265.2 | 7 | 1616 | ||
NM_153490.2 | NP_705694.2 | 8 | 1642 | |||
GenBank mRNA | AK092276.1 | - | 10 | 1571 | ||
AK223051.1 | - | 8 | 1641 | |||
AK223077.1 | - | 8 | 1641 | |||
BC002661.2 | - | 8 | 1626 | 3610361 | ||
BC077718.1 | - | 8 | 1619 | 5479174 | ||
X14640.1 | - | 8 | 1620 | |||
X52426.1 | - | 7 | 1600 |