Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175888.3 | NP_787084.1 | 4 | 400 | ||
XM_005265011.3 | XP_005265068.1 | 4 | 3662 | |||
XM_005265012.3 | XP_005265069.1 | 4 | 3587 | |||
XM_017006064.1 | XP_016861553.1 | 5 | 3799 | |||
XM_017006066.1 | XP_016861555.1 | 5 | 3703 | |||
XM_017006067.1 | XP_016861556.1 | 3 | 298 | |||
GenBank mRNA | AK131232.1 | - | 4 | 400 | ||
DA172830.1 | - | 4 | 400 |