Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177533.4 | NP_803877.2 | 5 | 788 | ||
GenBank mRNA | AB087802.1 | - | 5 | 674 | ||
AK128196.1 | - | 7 | 3253 | |||
BC041584.1 | - | 5 | 750 | 4810207 |