Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLX1A-SULT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037608.1 | 11 | 2180 | |||
GenBank mRNA | AK097386.1 | - | 8 | 1823 | ||
AK122707.1 | - | 11 | 2181 | |||
BC014471.1 | - | 9 | 1445 | 4907290 | ||
BC111011.1 | - | 10 | 2208 | 5768190 |
Gene Symbol: SULT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177552.3 | NP_808220.1 | 8 | 1363 | ||
GenBank mRNA | BC078144.1 | - | 8 | 1307 | 6022425 |