Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178129.4 | NP_835230.1 | 2 | 3179 | ||
XM_005274429.3 | XP_005274486.1 | 2 | 3613 | |||
XM_006724443.3 | XP_006724506.2 | 3 | 3692 | |||
XM_011546178.2 | XP_011544480.1 | 3 | 3296 | |||
XM_011546179.2 | XP_011544481.1 | 2 | 3256 | |||
GenBank mRNA | AK092956.1 | - | 1 | 1041 | ||
AL833184.1 | - | 1 | 1517 | |||
BC043610.1 | - | 2 | 3142 | 5787268 |