Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178229.4 | NP_839943.2 | 38 | 5912 | ||
XM_011509198.2 | XP_011507500.1 | 38 | 5901 | |||
XM_011509200.2 | XP_011507502.1 | 27 | 5081 | |||
XM_011509201.2 | XP_011507503.1 | 26 | 4470 | |||
XM_017000317.1 | XP_016855806.1 | 38 | 5898 | |||
XM_017000318.1 | XP_016855807.1 | 36 | 5655 | |||
GenBank mRNA | AB105103.1 | - | 38 | 5848 | ||
AK122653.1 | - | 24 | 4117 |