Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178450.4 | NP_848545.1 | 5 | 1182 | ||
XM_011543128.2 | XP_011541430.1 | 4 | 919 | |||
XM_011543129.1 | XP_011541431.1 | 6 | 1047 | |||
XM_011543130.1 | XP_011541432.1 | 6 | 952 | |||
XM_011543131.2 | XP_011541433.1 | 6 | 1181 | |||
XM_017009014.1 | XP_016864503.1 | 4 | 1370 | |||
GenBank mRNA | AF055007.1 | - | 5 | 838 | ||
AK290129.1 | - | 5 | 966 | |||
BC047569.1 | - | 5 | 932 | 4819831 |