Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT10 SIRNA Assays ›
Gene Symbol:
SEPT10
Gene Name:
septin 10
Gene Aliases:
-
Chromosome Location:
Chr.2: 109542797 - 109614247 on Build GRCh38
UniGene ID:
Hs.469615
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
290198
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321496.1
NP_001308425.1
9
2096
NM_001321498.1
NP_001308427.1
12
2493
NM_001321499.1
NP_001308428.1
7
3634
NM_001321500.1
NP_001308429.1
9
4066
NM_001321501.1
NP_001308430.1
10
4139
NM_001321502.1
NP_001308431.1
8
3933
NM_001321503.1
NP_001308432.1
5
3432
NM_001321504.1
NP_001308433.1
7
3752
NM_001321505.1
NP_001308434.1
8
2027
NM_001321506.1
NP_001308435.1
7
1840
NM_001321507.1
NP_001308436.1
8
3821
NM_001321508.1
NP_001308437.1
6
3619
NM_001321509.1
NP_001308438.1
11
2414
NM_001321510.1
NP_001308439.1
8
1909
NM_001321511.1
NP_001308440.1
6
3565
NM_001321512.1
NP_001308441.1
10
4135
NM_001321513.1
NP_001308442.1
10
4272
NM_001321514.1
NP_001308443.1
11
2238
NM_001321515.1
NP_001308444.1
10
2478
NM_144710.4
NP_653311.1
11
2410
NM_178584.3
NP_848699.1
10
2341
XM_011510698.2
XP_011509000.1
12
2326
XM_011510699.2
XP_011509001.1
11
2145
XM_011510700.2
XP_011509002.1
12
2496
XM_011510701.2
XP_011509003.1
10
2220
XM_011510702.2
XP_011509004.1
9
1991
XM_011510703.2
XP_011509005.1
12
2103
XM_011510704.2
XP_011509006.1
9
4002
GenBank mRNA
AB208875.1
-
6
3229
AF146760.1
-
11
2159
AK022682.1
-
8
1742
AK092033.1
-
10
2061
AK125706.1
-
1
1870
AL049934.1
-
5
1212
AL353938.1
-
6
1372
BC020502.1
-
11
2104
3913356
BC050345.1
-
12
2431
4830029
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle
cell division
protein binding
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0