Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313941.1 | NP_001300870.1 | 3, 4 | 539 | ||
NM_001313943.1 | NP_001300872.1 | 2, 3 | 444 | |||
NM_001313945.1 | NP_001300874.1 | 2, 3 | 321 | |||
NM_001664.3 | NP_001655.1 | 2, 3 | 444 | |||
GenBank mRNA | AF498970.1 | - | 1, 2 | 164 | ||
AK130066.1 | - | 2, 3 | 440 | |||
AK130808.1 | - | 2, 3 | 320 | |||
AK222556.1 | - | 2, 3 | 336 | |||
AK307220.1 | - | 2, 3 | 219 | |||
BC001360.2 | - | 2, 3 | 276 | 3051460 | ||
BF966305.1 | - | 3, 4 | 414 | |||
BQ721697.1 | - | 2, 3 | 317 | |||
BT019870.1 | - | 1, 2 | 164 | |||
BX647063.1 | - | 2, 3 | 340 | |||
L09159.1 | - | 1, 2 | 153 | |||
L25080.1 | - | 2, 3 | 315 | |||
X05026.1 | - | 2, 3 | 322 |