Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181503.2 | NP_852480.1 | 7 | 639 | ||
XM_006719763.1 | XP_006719826.1 | 6 | 770 | |||
GenBank mRNA | AF025438.1 | - | 6 | 368 | ||
AF279901.1 | - | 8 | 430 | |||
AK096810.1 | - | 5 | 2279 | |||
AK126602.1 | - | 1 | 824 | |||
AL050353.1 | - | 6 | 1862 | |||
BC020773.1 | - | 7 | 400 | 4808747 |