Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005809.5 | NP_005800.3 | 2 | 265 | ||
GenBank mRNA | AK022395.1 | - | 1 | 675 | ||
AK289485.1 | - | 2 | 185 | |||
AK293957.1 | - | 2 | 185 | |||
AL600939.1 | - | 2 | 155 | |||
BC000452.2 | - | 2 | 144 | 2821457 | ||
BC003022.1 | - | 2 | 144 | 2821457 | ||
BC039428.1 | - | 2 | 204 | 5303023 | ||
BC064138.1 | - | 2 | 144 | |||
BG705975.1 | - | 2 | 239 | |||
BP248561.1 | - | 2 | 265 | |||
CR450356.1 | - | 1 | 85 | |||
CR541789.1 | - | 1 | 85 | |||
EU668325.1 | - | 2 | 144 | |||
L19185.1 | - | 2 | 209 | |||
Z22548.1 | - | 2 | 174 |