Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021021.3 | NP_066301.1 | 6 | 1718 | ||
GenBank mRNA | AK026095.1 | - | 8 | 1861 | ||
AK292655.1 | - | 6 | 1792 | |||
BC098573.1 | - | 6 | 1678 | 30925462 |