Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCLAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002257.2 | NP_001002257.1 | 2 | 258 | ||
NM_001304445.1 | NP_001291374.1 | 4 | 663 | |||
NM_182551.4 | NP_872357.2 | 3 | 450 | |||
XM_005264244.1 | XP_005264301.1 | 3 | 412 | |||
XM_005264245.3 | XP_005264302.1 | 2 | 264 | |||
XM_011532741.2 | XP_011531043.1 | 3 | 401 | |||
XM_011532742.2 | XP_011531044.1 | 3 | 228 | |||
XM_011532743.2 | XP_011531045.1 | 2 | 215 | |||
XM_011532744.2 | XP_011531046.1 | 2 | 253 | |||
XM_011532745.1 | XP_011531047.1 | 3 | 412 | |||
XM_017003746.1 | XP_016859235.1 | 3 | 295 | |||
XM_017003747.1 | XP_016859236.1 | 3 | 412 | |||
GenBank mRNA | AK095284.1 | - | 3 | 415 | ||
AK296965.1 | - | 2 | 243 | |||
AK302562.1 | - | 4 | 616 | |||
AY358702.1 | - | 3 | 419 | |||
BC146817.1 | - | 2 | 248 | 8860419 | ||
BP872689.1 | - | 2 | 258 | |||
BX647953.1 | - | 2 | 214 |