Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278590.1 | NP_001265519.1 | 2 | 199 | ||
NM_001278591.1 | NP_001265520.1 | 2 | 199 | |||
NM_001278592.1 | NP_001265521.1 | 2 | 199 | |||
NM_006859.3 | NP_006850.2 | 2 | 199 | |||
NM_194451.2 | NP_919433.1 | 2 | 199 | |||
XM_006713990.3 | XP_006714053.1 | 2 | 201 | |||
XM_017007665.1 | XP_016863154.1 | 2 | 192 | |||
XM_017007666.1 | XP_016863155.1 | 2 | 201 | |||
XM_017007667.1 | XP_016863156.1 | 2 | 192 | |||
GenBank mRNA | AK026289.1 | - | 2 | 167 | ||
AK291985.1 | - | 2 | 130 | |||
AK292238.1 | - | 2 | 199 | |||
AK303429.1 | - | 2 | 178 | |||
AL528262.3 | - | 2 | 149 | |||
AV729111.1 | - | 2 | 184 | |||
BC023635.2 | - | 2 | 144 | 4841470 | ||
BC062751.1 | - | 2 | 169 |