Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021033.6 | NP_066361.1 | 2 | 2145 | ||
GenBank mRNA | AF205602.1 | - | 3 | 713 | ||
AK090533.1 | - | 1 | 90 | |||
AK094586.1 | - | 1 | 876 | |||
BC041333.2 | - | 2 | 2146 | 5278625 | ||
BC070031.1 | - | 2 | 1903 | 4815530 |