Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207009.1 | NP_001193938.1 | 2 | 877 | ||
NM_198458.2 | NP_940860.2 | 3 | 974 | |||
GenBank mRNA | AK126727.1 | - | 3 | 880 | ||
AK131209.1 | - | 2 | 877 | |||
BC118983.1 | - | 2 | 742 | 40025731 | ||
BC125273.1 | - | 2 | 747 | 40025732 |
Gene Symbol: LOC100419840 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC023200.1 | - | 3 | 1889 | ||
BC023201.1 | - | 3 | 1889 |