Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF615 SIRNA Assays ›
Gene Symbol:
ZNF615
Gene Name:
zinc finger protein 615
Gene Aliases:
-
Chromosome Location:
Chr.19: 51991332 - 52008230 on Build GRCh38
UniGene ID:
Hs.368355
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
290290
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF615
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199324.1
NP_001186253.1
5
528
NM_001321317.1
NP_001308246.1
3
286
NM_001321318.1
NP_001308247.1
4
324
NM_001321319.1
NP_001308248.1
4
490
NM_001321320.1
NP_001308249.1
4
490
NM_001321321.1
NP_001308250.1
4
483
NM_001321322.1
NP_001308251.1
4
490
NM_001321323.1
NP_001308252.1
3
286
NM_198480.3
NP_940882.3
5
528
XM_011526824.1
XP_011525126.1
6
806
XM_011526825.1
XP_011525127.1
6
932
XM_011526826.1
XP_011525128.1
5
772
XM_011526829.1
XP_011525131.1
4
454
XM_011526830.1
XP_011525132.1
3
257
XM_017026648.1
XP_016882137.1
6
806
XM_017026649.1
XP_016882138.1
6
932
XM_017026650.1
XP_016882139.1
5
519
XM_017026651.1
XP_016882140.1
6
932
GenBank mRNA
AK128361.1
-
4
294
AK131494.1
-
5
503
AK291734.1
-
5
501
AK294977.1
-
3
286
BC053884.1
-
4
476
BC105100.1
-
3
249
8144103
BC105102.1
-
3
249
8144105
BC112961.1
-
4
437
BC143429.1
-
3
249
9051937
BX647556.1
-
4
456
CR936612.1
-
5
508
DA095942.1
-
5
528
DA119086.1
-
4
493
DA236166.1
-
5
501
DA240372.1
-
3
286
DA244381.1
-
5
530
DA322276.1
-
4
488
DA802380.1
-
4
454
DB235427.1
-
4
463
DB290987.1
-
4
451
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0