Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF621 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098414.2 | NP_001091884.1 | 5 | 600 | ||
NM_001287245.1 | NP_001274174.1 | 5 | 836 | |||
NM_198484.4 | NP_940886.1 | 5 | 836 | |||
XM_005265079.3 | XP_005265136.1 | 5 | 663 | |||
XM_005265080.3 | XP_005265137.1 | 4 | 582 | |||
XM_006713114.3 | XP_006713177.1 | 5 | 544 | |||
XM_011533610.2 | XP_011531912.1 | 5 | 1170 | |||
XM_017006226.1 | XP_016861715.1 | 4 | 463 | |||
GenBank mRNA | AK074366.1 | - | 5 | 761 | ||
AK127181.1 | - | 5 | 600 | |||
BC101619.1 | - | 5 | 474 | 8069125 | ||
BC113415.1 | - | 5 | 474 | 8322467 | ||
CR749268.1 | - | 5 | 701 |