Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198946.2 | NP_945184.1 | 2 | 188 | ||
GenBank mRNA | AF303084.1 | - | 2 | 185 | ||
AY301269.1 | - | 2 | 143 | |||
AY358598.1 | - | 2 | 168 | |||
BC062746.1 | - | 2 | 189 | 6452008 |