Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199243.1 | NP_954713.1 | 1 | 242 | ||
GenBank mRNA | BC030197.1 | - | 1 | 440 | ||
JF810884.1 | - | 1 | 242 |
Set Membership: |
G-protein coupled receptor |