Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GINS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021067.4 | NP_066545.3 | 7 | 1507 | ||
NR_134574.1 | 8 | 1629 | ||||
XM_017028162.1 | XP_016883651.1 | 7 | 1464 | |||
GenBank mRNA | BC012542.1 | - | 7 | 1508 | 4333095 | |
D80008.1 | - | 7 | 1462 |