Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBPJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005349.3 | NP_005340.2 | 5 | 575 | ||
NM_015874.4 | NP_056958.3 | 4 | 522 | |||
NM_203283.2 | NP_976028.1 | 4 | 459 | |||
NM_203284.2 | NP_976029.1 | 5 | 705 | |||
XM_005248161.3 | XP_005248218.1 | 5 | 589 | |||
XM_011513840.2 | XP_011512142.1 | 5 | 981 | |||
XM_017008170.1 | XP_016863659.1 | 4 | 378 | |||
XM_017008171.1 | XP_016863660.1 | 4 | 357 | |||
XM_017008172.1 | XP_016863661.1 | 5 | 520 | |||
XM_017008173.1 | XP_016863662.1 | 5 | 798 | |||
XM_017008174.1 | XP_016863663.1 | 5 | 614 | |||
XM_017008175.1 | XP_016863664.1 | 5 | 530 | |||
GenBank mRNA | AK302230.1 | - | 4 | 312 | ||
AK303244.1 | - | 4 | 371 | |||
AK311419.1 | - | 4 | 358 | |||
BC020780.1 | - | 4 | 459 | 4718839 | ||
BC064976.1 | - | 5 | 480 | 6152924 | ||
BE269143.1 | - | 5 | 486 | |||
BM472788.1 | - | 529 | ||||
BM555733.1 | - | 5 | 683 | |||
BQ216033.1 | - | 4 | 503 | |||
D14041.1 | - | 2 | 352 | |||
DC361185.1 | - | 4 | 523 | |||
L07872.1 | - | 4 | 417 |