Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078471.3 | NP_510880.2 | 2 | 751 | ||
NM_203318.1 | NP_976063.1 | 2 | 751 | |||
GenBank mRNA | AB177858.1 | - | 1 | 570 | ||
BC039612.1 | - | 2 | 855 | 6045482 | ||
CR933614.1 | - | 2 | 726 | |||
D86970.2 | - | 2 | 774 |