Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021083.2 | NP_066569.1 | 1, 2 | 335 | ||
XM_011543978.2 | XP_011542280.1 | 1, 2 | 449 | |||
GenBank mRNA | AY534238.1 | - | 1, 2 | 459 | ||
BC036019.1 | - | 1, 2 | 376 | 5272840 | ||
Z32684.2 | - | 1, 2 | 335 |