Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105378260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028520.1 | XP_016884009.1 | 7 | 1307 | ||
XM_017028521.1 | XP_016884010.1 | 7 | 1307 |
Gene Symbol: BAGE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182482.2 | NP_872288.2 | 10 | 1856 | ||
GenBank mRNA | AF218570.1 | - | 10 | 1856 |