Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAHD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320848.1 | NP_001307777.1 | 7 | 1062 | ||
NM_199336.2 | NP_955368.1 | 6 | 936 | |||
XM_011510745.2 | XP_011509047.1 | 7 | 1052 | |||
XM_011510746.2 | XP_011509048.1 | 7 | 1171 | |||
XM_011510747.2 | XP_011509049.1 | 6 | 936 | |||
XM_011510748.2 | XP_011509050.1 | 7 | 1052 | |||
XM_011510750.2 | XP_011509052.1 | 5 | 831 | |||
XM_017003470.1 | XP_016858959.1 | 7 | 1049 | |||
XM_017003471.1 | XP_016858960.1 | 7 | 1052 | |||
XM_017003472.1 | XP_016858961.1 | 6 | 985 | |||
XM_017003473.1 | XP_016858962.1 | 4 | 713 | |||
GenBank mRNA | AL833869.1 | - | 5 | 736 | ||
BC064511.1 | - | 6 | 899 | 6644987 |
Gene Symbol: FAHD2CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003698.1 | 2 | 245 | |||
GenBank mRNA | BI916267.1 | - | 2 | 212 | ||
BM790702.1 | - | 2 | 245 | |||
CN274361.1 | - | 2 | 178 |