Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144895.1 | NP_001138367.1 | 2 | 163 | ||
NM_001144896.1 | NP_001138368.1 | 2 | 163 | |||
NM_001144897.1 | NP_001138369.1 | 2 | 163 | |||
NM_001144899.1 | NP_001138371.1 | 2 | 163 | |||
NM_021155.3 | NP_066978.1 | 2 | 163 | |||
NR_026692.1 | 1 | 263 | ||||
XM_005272472.4 | XP_005272529.1 | 2 | 194 | |||
GenBank mRNA | AF290886.1 | - | 2 | 105 | ||
AK293089.1 | - | 2 | 119 | |||
AK304190.1 | - | 2 | 149 | |||
AK313585.1 | - | 2 | 164 | |||
AY042221.1 | - | 2 | 96 | |||
AY042222.1 | - | 2 | 96 | |||
AY042223.1 | - | 2 | 96 | |||
AY042224.1 | - | 2 | 96 | |||
AY042225.1 | - | 2 | 96 | |||
AY042229.1 | - | 1 | 196 | |||
AY042230.1 | - | 1 | 196 | |||
AY042231.1 | - | 1 | 196 | |||
AY042232.1 | - | 1 | 196 | |||
AY042233.1 | - | 1 | 196 | |||
BC110615.2 | - | 1 | 203 | 40032459 | ||
M98457.1 | - | 2 | 137 |
Gene Symbol: CLEC4M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC110614.2 | - | 105 | 40032453 |