Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRINP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021165.3 | NP_066988.1 | 2 | 636 | ||
XM_005245379.1 | XP_005245436.1 | 3 | 1216 | |||
GenBank mRNA | AB051534.1 | - | 2 | 629 | ||
AB161694.1 | - | 2 | 629 | |||
AK296524.1 | - | 2 | 592 | |||
AL035289.1 | - | 1 | 210 | |||
BC028036.1 | - | 2 | 528 | 5242899 |