Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021178.4 | NP_067001.3 | 5 | 707 | ||
NM_182849.2 | NP_878269.1 | 4 | 507 | |||
NM_182852.3 | NP_878272.1 | 6 | 888 | |||
GenBank mRNA | AF216381.1 | - | 1 | 218 | ||
AK026233.1 | - | 5 | 674 | |||
AK225253.1 | - | 5 | 679 | |||
AK225451.1 | - | 5 | 674 | |||
AK226037.1 | - | 5 | 679 | |||
AK309183.1 | - | 5 | 676 | |||
AL161994.1 | - | 5 | 687 | |||
BC000369.2 | - | 4 | 649 | 2819346 | ||
BC001218.1 | - | 4 | 652 | 3528496 | ||
BC004435.1 | - | 4 | 649 | 2819346 | ||
BI771433.1 | - | 5 | 690 | |||
BI917081.1 | - | 5 | 698 | |||
BM741413.1 | - | 4 | 474 | |||
BX161423.1 | - | 5 | 867 | |||
BX161424.1 | - | 5 | 835 | |||
BX247978.1 | - | 5 | 815 | |||
BX334446.2 | - | 5 | 830 |