Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001868.3 | NP_001859.1 | 4 | 581 | ||
GenBank mRNA | AK131054.1 | - | 916 | |||
AK291493.1 | - | 4 | 458 | |||
AK293366.1 | - | 4 | 393 | |||
BC005279.1 | - | 4 | 453 | 3949850 | ||
BT007313.1 | - | 4 | 431 | |||
DC378103.1 | - | 4 | 581 | |||
X67318.1 | - | 4 | 438 |