Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130851.1 | NP_001124323.1 | 6 | 492 | ||
NM_005192.3 | NP_005183.2 | 7 | 612 | |||
XM_011536353.2 | XP_011534655.1 | 7 | 628 | |||
GenBank mRNA | AF213033.1 | - | 6 | 498 | ||
AF213036.1 | - | 6 | 498 | |||
AF213038.1 | - | 5 | 378 | |||
AF213039.1 | - | 6 | 498 | |||
AF213041.1 | - | 6 | 498 | |||
AF213042.1 | - | 5 | 378 | |||
AF213044.1 | - | 4 | 360 | |||
AF213045.1 | - | 6 | 480 | |||
AF213046.1 | - | 6 | 498 | |||
AF213047.1 | - | 5 | 378 | |||
AF213048.1 | - | 4 | 360 | |||
AF213049.1 | - | 5 | 378 | |||
AF213050.1 | - | 6 | 483 | |||
AF213053.1 | - | 6 | 498 | |||
AK098653.1 | - | 7 | 612 | |||
AY257474.1 | - | 6 | 498 | |||
AY257475.1 | - | 3 | 198 | |||
BC064965.1 | - | 6 | 452 | 6143626 | ||
BT019451.1 | - | 6 | 498 | |||
CR407666.1 | - | 6 | 498 | |||
EF560750.1 | - | 7 | 538 | |||
KP966095.1 | - | 6 | 529 | |||
L25876.1 | - | 7 | 559 | |||
L27711.1 | - | 7 | 549 | |||
U02681.1 | - | 7 | 529 |