Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005632.2 | NP_005623.1 | 14 | 4599 | ||
XM_011522620.2 | XP_011520922.1 | 15 | 7922 | |||
XM_011522621.2 | XP_011520923.1 | 15 | 8850 | |||
XM_011522622.1 | XP_011520924.1 | 13 | 7554 | |||
XM_011522623.2 | XP_011520925.1 | 16 | 5625 | |||
XM_011522624.2 | XP_011520926.1 | 15 | 4963 | |||
XM_011522625.2 | XP_011520927.1 | 14 | 4849 | |||
XM_011522626.2 | XP_011520928.1 | 15 | 8390 | |||
XM_011522627.2 | XP_011520929.1 | 15 | 8391 | |||
XM_011522628.2 | XP_011520930.1 | 15 | 5483 | |||
XM_011522629.2 | XP_011520931.1 | 15 | 5346 | |||
XM_011522630.2 | XP_011520932.1 | 14 | 4684 | |||
XM_011522631.2 | XP_011520933.1 | 13 | 4570 | |||
XM_011522632.2 | XP_011520934.1 | 13 | 5001 | |||
XM_017023596.1 | XP_016879085.1 | 16 | 5762 | |||
GenBank mRNA | BC001869.1 | - | 2 | 1325 | ||
BC021854.1 | - | 10 | 2666 | |||
BC040169.1 | - | 1 | 795 | |||
BC112918.1 | - | 6 | 2824 | |||
U69557.1 | - | 2 | 1285 |