Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007271.1 | NP_001007272.1 | 8 | 1598 | ||
NM_001007272.1 | NP_001007273.1 | 4 | 873 | |||
NM_001007273.1 | NP_001007274.1 | 6 | 1172 | |||
NM_001320842.1 | NP_001307771.1 | 7 | 1390 | |||
NM_001320843.1 | NP_001307772.1 | 5 | 968 | |||
NM_016364.3 | NP_057448.3 | 4 | 731 | |||
XM_005269883.3 | XP_005269940.1 | 7 | 1368 | |||
XM_005269884.4 | XP_005269941.3 | 4 | 1160 | |||
XM_005269887.1 | XP_005269944.1 | 7 | 1470 | |||
XM_005269890.1 | XP_005269947.1 | 5 | 1070 | |||
XM_011539853.1 | XP_011538155.1 | 8 | 1607 | |||
XM_011539854.2 | XP_011538156.1 | 6 | 1146 | |||
XM_011539855.1 | XP_011538157.1 | 4 | 849 | |||
XM_011539856.2 | XP_011538158.1 | 6 | 1150 | |||
XM_017016313.1 | XP_016871802.1 | 4 | 823 | |||
GenBank mRNA | AB027004.1 | - | 4 | 745 | ||
AB103375.1 | - | 4 | 745 | |||
AK057012.1 | - | 5 | 941 | |||
AK127886.1 | - | 7 | 1390 | |||
AY040091.1 | - | 8 | 1562 | |||
AY674051.1 | - | 8 | 1571 | |||
BC009778.1 | - | 5 | 857 | 4047963 |