Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077238.1 | NP_001070706.1 | 3 | 384 | ||
NM_152988.2 | NP_694533.1 | 3 | 384 | |||
XM_011528138.2 | XP_011526440.1 | 3 | 443 | |||
XM_011528139.1 | XP_011526441.1 | 2 | 238 | |||
XM_017027016.1 | XP_016882505.1 | 3 | 443 | |||
XM_017027017.1 | XP_016882506.1 | 4 | 574 | |||
XM_017027018.1 | XP_016882507.1 | 2 | 238 | |||
XM_017027019.1 | XP_016882508.1 | 4 | 574 | |||
GenBank mRNA | AB040965.1 | - | 3 | 333 | ||
AJ345027.1 | - | 3 | 337 | |||
AJ420897.1 | - | 3 | 333 | |||
AK308973.1 | - | 4 | 453 | |||
AK309122.1 | - | 3 | 458 | |||
AY169315.1 | - | 3 | 332 | |||
BC001788.1 | - | 3 | 318 | 3352341 | ||
BC028391.2 | - | 3 | 321 | 4822524 | ||
BC093046.1 | - | 3 | 384 | 30395602 |