Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242924.1 | NP_001229853.1 | 6 | 935 | ||
NM_001242926.1 | NP_001229855.1 | 5 | 884 | |||
NM_001242927.1 | NP_001229856.1 | 4 | 665 | |||
NM_001242928.1 | NP_001229857.1 | 5 | 884 | |||
NM_021188.2 | NP_067011.1 | 5 | 884 | |||
NR_040251.1 | 5 | 852 | ||||
GenBank mRNA | AK223261.1 | - | 5 | 640 | ||
AK293350.1 | - | 6 | 691 | |||
AK298307.1 | - | 6 | 607 | |||
AK298520.1 | - | 5 | 660 | |||
AK299138.1 | - | 4 | 663 | |||
AK301382.1 | - | 5 | 537 | |||
AK303834.1 | - | 5 | 550 | |||
AK309957.1 | - | 6 | 680 | |||
AK310031.1 | - | 4 | 426 | |||
AK315206.1 | - | 5 | 642 | |||
BC004357.1 | - | 5 | 628 | |||
BC050683.1 | - | 5 | 694 | 5752452 | ||
U90919.1 | - | 5 | 616 |