Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CADM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021189.3 | NP_067012.1 | 2 | 292 | ||
GenBank mRNA | AF062733.2 | - | 2 | 249 | ||
AK289541.1 | - | 2 | 292 | |||
AK308915.1 | - | 2 | 264 |